Microscope Slides, Regular Non-Charged, 45° Corners
Supplier: Mortech Manufacturing
SL045-YL-BX SL045-BL-BX SL045-BL-CS SL045-GR-BX SL045-GR-CS SL045-LI-BX SL045-LI-CS SL045-PE-BX SL045-PE-CS SL045-PK-BX SL045-PK-CS SL045-TN-BX SL045-TN-CS SL045-WH-BX SL045-WH-CS SL045-YL-CS
76948-318BX 36.79 USD
76948-318 76948-290 76948-292 76948-294 76948-296 76948-298 76948-300 76948-302 76948-304 76948-306 76948-308 76948-310 76948-312 76948-314 76948-316 76948-320
Microscope Slides, Regular Non-Charged, 45° Corners
Slides Microscope Slides
High-quality, uncharged microscope slides with color coding.
  • 1×3" slides, 1 mm thickness
  • 100% guaranteed against sticking and contamination
  • 45 degree corners
  • Uniform flat paint for perfect barcodes
Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac.
Packaging: The packaging options include 1 box of 72 slides and 1 case of 1440 slides.