Coverslips
Supplier: Mopec
SL024-50-10 SL024-50-20 SL024-55-10 SL024-55-20 SL024-60-10
76948-280CS 174.91 USD
76948-280 76948-282 76948-284 76948-286 76948-288
Coverslips
Cover Glasses
Mopec coverslips are made with borosilicate glass.
  • Borosilicate glass, #1 thickness, 12 mm in diameter
  • Will not stick or auto-fluoresce
  • Clean, uniform surface
  • Ideal for automated coverslipping
Available in 1 or 2 ounce volumes.
Packaging: The coverslips are packaged 10 boxes per case, with 100 or 200 pieces per box.