Control Slides, Tissue Microarray, Breast Cancer and Normal Adjacent Tissue, 2 Matched Pairs
Supplier: Mortech Manufacturing
SC988-25
76948-272CS 2004.92 USD
76948-272
Control Slides, Tissue Microarray, Breast Cancer and Normal Adjacent Tissue, 2 Matched Pairs
Slides Microscopy Control Slides
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Packaging: Case of 25 contains 24 unstained slides and 1 stained slide.