PIP® Corded Ear Plugs
Supplier: PIP
267-HPR400C
76944-646BX 67.96 USD
76944-646
PIP® Corded Ear Plugs
Earplugs Reusable Earplugs
Reusable TPR corded ear plugs provide NRR 27dB protection.
  • Pre-formed plug shape with no rolling required makes insertion simple
  • Self-sealing flanges give all day comfort
  • Rigid stem makes insertion easy and reduces the risk of contamination
  • Non-resonating vinyl cord for comfort and easy storage
  • Includes removable cord
Self-sealing, 4-flange TPR (thermoplastic rubber) construction for all-day use and secure sound attenuation. The TPR plug can be cleaned using soap and water, and can be reused several times, making them very cost-efficient. Moderate attenuation allows for safe usage that blocks out hazardous noise yet allows warning signals and voices to be clearly understood.