PIP® EZ-Twist™ Polyurethane Foam Corded Ear Plugs
Supplier: PIP
267-HPF610C
76944-642BX 54.2 USD
76944-642
PIP® EZ-Twist™ Polyurethane Foam Corded Ear Plugs
Earplugs Disposable Earplugs
Disposable soft polyurethane foam corded ear plugs provide NRR 30dB protection.
  • Just insert and twist
  • Fast fitting
  • Shape conforms to most ear canals
  • 76944-642 cylinder shap
  • Black PVC cord for convenience
Hybrid foam plug and unique stem enable easy insertion, with no need to roll down before fitting. Ideal for use with dirty hands. The PowerSoft™ Foam Technology features a proprietary polyurethane formulation that delivers optimal recovery time, easy insertion, and maximum all-day comfort. Shape conforms to most ear canals for maximum sound attenuation.