PIP® Bouton® Universal Fit PETG Safety Visor
Supplier: PIP
251-01-5211
76944-636CS 179.92 USD
76944-636
PIP® Bouton® Universal Fit PETG Safety Visor
Face Shields
Universal Fit PETG (Polyethylene Terephthalate Glycol) Safety Visor with 0.040" thick lens.
  • Made with virgin resin for better uniformity of the viewing area
  • Universal hole pattern will fit most headgear and hard hat adapters
  • Ships with masking on both sides to prevent scratching
PETG (Polyethylene Terephthalate Glycol) safety visor is ideal for liquid splash applications.