PIP® Bouton® Universal Fit Polycarbonate Safety Visor with Chin Cup
Supplier: PIP
251-01-5210
76944-634CS 266.42 USD
76944-634
PIP® Bouton® Universal Fit Polycarbonate Safety Visor with Chin Cup
Face Shields
Universal fit polycarbonate safety visor with chin cup features an 0.040" thick lens.
  • Clear polycarbonate visor
  • Pre-assembled clear chin cup provides additional protection
  • Universal hole pattern will fit most headgear and hard hat adapters
  • Ships with masking on both sides to prevent scratching
Polycarbonate safety visor with aluminum binding is ideal for high impact applications. Outperforms PETG (Polyethylene Terephthalate Glycol) in a wider range of temperatures.
Certifications: Meets ANSI Z87.1+ requirements.