PIP® Anser™ Semi-Rimless Safety Readers with +1.50 Diopter
Supplier: PIP
250-AN-52015
76944-630CS 617.7 USD
76944-630
PIP® Anser™ Semi-Rimless Safety Readers with +1.50 Diopter
Safety Glasses
Semi-rimless safety readers feature a black frame, clear lens, +1.50 diopter, and anti-scratch Fogless® 3Sixty™ coating.
  • Magnification reader glasses are for the worker who needs to see up close
  • Lightweight, semi-rimless design provides all day user comfort
  • Bayonet temples with flexible rubber tips offer a comfortable, secure fit
  • Soft rubber nose pads help keep the glasses in place
  • Supplied with an adjustable neck cord
  • 8.5 Base curve lens
Sleek, sporty style with wrap-around lens helps provide protection from projectiles, impact, and liquid splash. The FogLess® 3Sixty™ enhanced hydrophobic coating resists lens fogging and provides clear vision in extreme working conditions.
Certifications: Meets ANSI Z87.1-2015 standards.