PIP® Recon™ Rimless Safety Glasses
Supplier: PIP
250-32-0520
76944-626CS 931.93 USD
76944-626
PIP® Recon™ Rimless Safety Glasses
Safety Glasses
Rimless safety glasses feature a clear temple, clear lens, and anti-scratch FogLess® 3Sixty™ coating.
  • Lightweight, rimless design provides all day comfort
  • Flexible bayonet temples contain rubber pads for a secure, non-slip fit
  • Adjustable rubber nosepiece provides a comfortable fit
  • 9 Base curve lens
Lightweight, rimless design provides an ergonomic fit, with flexible bayonet temples that contain rubber pads for an improved fit. The FogLess® 3Sixty™ enhanced hydrophobic coating resists lens fogging and provides clear vision in extreme working conditions.
Certifications: Meets ANSI Z87.1+. Meets CSA Z94.3.