NOVAX® Straight Cuff Rubber Insulating Gloves, Black/Orange
Supplier: PIP
155-0-11/10 155-0-11/11 155-0-11/12 155-0-11/7 155-0-11/8 155-0-11/9
76944-524PR 86.33 USD
76944-524 76944-526 76944-528 76944-518 76944-520 76944-522
NOVAX® Straight Cuff Rubber Insulating Gloves, Black/Orange
Gloves
Class 0 black/orange rubber insulating gloves with a straight cuff, available in an 11" length.
  • Contoured shape reduces hand fatigue
  • Silicone-free
  • Individually numbered for traceability and stamped with factory manufacturing date
Natural rubber construction offers the required dielectric properties, combined with flexibility, strength, and durability.
Certifications: Gloves meet ANSI/ASTM D120 standard and NFPA 70E for use around electrical hazards and arc flash protection.