Methyl 3-bromo-2-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

50668-1G 50668-25G 50668-500MG 50668-5G
76942-948EA 137.92 USD
76942-948 76942-950 76942-952 76942-954
Methyl 3-bromo-2-iodobenzoate ≥97%
Methyl 3-bromo-2-iodobenzoate

Caution: for Research Use Only

Formula: C₈H₆BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD18398801
CAS Number: 121772-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR