5-Methoxynicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

14151-25G
76936-848EA 148.53 USD
76936-848
5-Methoxynicotinaldehyde ≥95%
5-Methoxynicotinaldehyde
Formula: C7H7NO2
Storage Temperature: Ambient
MDL Number: MFCD07437946
CAS Number: 113118-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR