Potassium isopropyltrifluoroborate ≥97%

Supplier: AOBChem USA

Warning

12424-25G
76936-588EA 169.75 USD
76936-588
Potassium isopropyltrifluoroborate ≥97%
Potassium isopropyltrifluoroborate
Formula: C₃H₇BF₃K
Storage Temperature: Ambient
MDL Number: MFCD04112718
CAS Number: 1041642-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR