N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥95% research grade

Supplier: AOBChem USA

Warning

10097-25G
76936-336EA 133.68 USD
76936-336
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥95% research grade
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
Formula: C₁₆H₂₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00060156
CAS Number: 29473-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR