5-Bromo-2-fluoroisonicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

31706-1G 31706-25G 31706-5G
76930-718EA 117.76 USD
76930-718 76930-720 76930-722
5-Bromo-2-fluoroisonicotinic acid ≥97% research grade
5-Bromo-2-fluoroisonicotinic acid
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185803
CAS Number: 1214377-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR