4-Bromo-2-methoxythiazole ≥97% research grade

Supplier: AOBChem USA

Warning

26922-1G 26922-25G 26922-5G
76929-346EA 66.84 USD
76929-346 76929-348 76929-350
4-Bromo-2-methoxythiazole ≥97% research grade
4-Bromo-2-methoxythiazole
Formula: C₄H₄BrNOS
Storage Temperature: Ambient
MDL Number: MFCD07787374
CAS Number: 240816-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR