4-Bromo-2-methoxythiazole ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
26922-1G
26922-25G
26922-5G
76929-346EA
66.84
USD
76929-346
76929-348
76929-350
4-Bromo-2-methoxythiazole ≥97% research grade
4-Bromo-2-methoxythiazole
Formula:
C₄H₄BrNOS Storage Temperature: Ambient |
MDL Number:
MFCD07787374 CAS Number: 240816-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...