4-Methoxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12152-25G
76924-698EA 127.31 USD
76924-698
4-Methoxy-2-methylphenylboronic acid ≥97%
4-Methoxy-2-methylphenylboronic acid

Caution: for Research Use Only

Formula: C₈H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD02684315
CAS Number: 208399-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR