4-Hexylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

12105-25G 12105-5G
76924-676EA 357.53 USD
76924-676 76924-678
4-Hexylphenylboronic acid ≥95%
4-Hexylphenylboronic acid

Caution: for Research Use Only

Formula: C₁₂H₁₉BO₂
Storage Temperature: Ambient
MDL Number: MFCD04039029
CAS Number: 105365-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR