5-Bromo-2-methylanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-methoxy-1-methylbenzene

10429-25G
76924-312EA 98.67 USD
76924-312
5-Bromo-2-methylanisole ≥97%
5-Bromo-2-methylanisole

Caution: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797972
CAS Number: 67868-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR