4-(tert-Butoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10356-25G
76924-296EA 112.46 USD
76924-296
4-(tert-Butoxycarbonyl)phenylboronic acid ≥97%
4-(tert-Butoxycarbonyl)phenylboronic acid

Caution: for Research Use Only

Formula: C₁₁H₁₅BO₄
Storage Temperature: Ambient
MDL Number: MFCD03411946
CAS Number: 850568-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR