4-Carboxy-3-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-3-chlorobenzeneboronic acid

10205-25G
76924-264EA 112.46 USD
76924-264
4-Carboxy-3-chlorophenylboronic acid ≥97%
4-Carboxy-3-chlorophenylboronic acid

Caution: for Research Use Only

Formula: C₇H₆BClO₄
MW: 200.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656268
CAS Number: 136496-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR