4-Chloro-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-methylbenzeneboronic acid

10185-25G
76924-252EA 62.6 USD
76924-252
4-Chloro-2-methylphenylboronic acid ≥97%
4-Chloro-2-methylphenylboronic acid

Caution: for Research Use Only

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683107
CAS Number: 209919-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR