4-Chloro-2-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane

10075-25G
76924-202EA 66.84 USD
76924-202
4-Chloro-2-(trifluoromethyl)phenylboronic acid ≥97%
4-Chloro-2-(trifluoromethyl)phenylboronic acid

Caution: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973086
CAS Number: 313545-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR