5-Chloro-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloropyridine-3-boronic acid, (5-Chloropyridin-3-yl)boronic acid

10043-25G
76924-190EA 88.06 USD
76924-190
5-Chloro-3-pyridineboronic acid ≥97%
5-Chloro-3-pyridineboronic acid

Caution: for Research Use Only

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798243
CAS Number: 872041-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR