3,5-Dibromobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16483-25G
76908-536EA 77.45 USD
76908-536
3,5-Dibromobenzaldehyde ≥97%
3,5-Dibromobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₄Br₂O
Storage Temperature: Ambient
MDL Number: MFCD00156887
CAS Number: 56990-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR