3-Bromophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10307-25G
76906-354EA 62.6 USD
76906-354
3-Bromophenylboronic acid ≥97%
3-Bromophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD00239386
CAS Number: 89598-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR