BD MAX™ Swab Collection Kit
Supplier: BD
443925
76906-236PK 325.63 USD
76906-236
BD MAX™ Swab Collection Kit
Swabs Swabs for Clinical Use
BD MAX™swab collection kit.
  • Multiple BD assays from one swab
  • Eliminates re-capping step by lab technician and helps to reduce waste (1 cap/sample)
  • Post-testing, a sample buffer tube with a pierced cap may also be stored upright
  • for up to 4 days at 2 to 30 °C
  • Up to 4x longer transport stability vs. UVE specimen collection Kit
  • for BD CTGCTV2 for BD MAX™ (21 days vs. 5 days)
  • Elimination of Pre-warm step for swab samples