BD MAX™ CTGCTV2 Kit
Supplier: BD
443904
76906-216EA 1298.46 USD
76906-216
BD MAX™ CTGCTV2 Kit
Clinical Diagnostic Supplies Clinical Reagents and Kits
The BD MAX™ CTGCTV2 test simultaneously (CT), Neisseria gonorrhoeae (GC) and Trichomonas vaginalis (TV).
  • Chlamydia trachomatis
  • Neisseria gonorrhoeae
  • Trichomonas vaginalis
  • Molecular
The BD MAX™ CTGCTV2 test elevates the standard of testing by addressing the three most prevalent non-viral sexually transmitted infections (STIs) in one assay.