BD MAX™ GBS Kit
Supplier: BD
441772
76906-208EA 1724.82 USD
76906-208
BD MAX™ GBS Kit
Clinical Diagnostic Supplies Clinical Reagents and Kits
The BD MAX™ GBS assay provides timely and accurate Group B Streptococcus screening results to help reduce the risk of GBS disease in newborn babies
  • Group B Streptococcus
  • Detects GBS DNA in Lim Brothcultures from antepartum women
  • Molecular
The BD MAX™ GBS assay, run on the fully-automated,easy-to-use BD MAX™ System, detects GBS DNA in Lim Broth cultures from antepartum women.4 The BD accommodates on-demand and batch workflows to meet MAX™ System your laboratory’s needs.