2-(Difluoromethoxy)-5-fluorobromobenzene ≥97%

Supplier: AOBChem USA

Warning

36369-1G 36369-25G 36369-500MG 36369-5G
76903-270EA 292.81 USD
76903-270 76903-272 76903-274 76903-276
2-(Difluoromethoxy)-5-fluorobromobenzene ≥97%
2-(Difluoromethoxy)-5-fluorobromobenzene
Formula: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09935620
CAS Number: 936249-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR