2,6-Dichloropyridine-4-methanol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine

26703-25G 26703-5G
76900-418EA 418 USD
76900-418 76900-420
2,6-Dichloropyridine-4-methanol ≥97% research grade
2,6-Dichloropyridine-4-methanol
Formula: C₆H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00052638
CAS Number: 101990-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR