2-Hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

22376-25G
76898-648EA 62.6 USD
76898-648
2-Hydroxyphenylboronic acid pinacol ester ≥97%
2-Hydroxyphenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₂H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD02093754
CAS Number: 269409-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR