2-Acetamido-5-bromopyridine ≥97%

Supplier: AOBChem USA

Warning

20846-25G
76898-194EA 70.02 USD
76898-194
2-Acetamido-5-bromopyridine ≥97%
2-Acetamido-5-bromopyridine

Caution: for Research Use Only

Formula: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD00468968
CAS Number: 7169-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR