2-Chloro-4-iodo-6-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

17434-25G
76896-848EA 141.1 USD
76896-848
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine ≥97%
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₃IN
Storage Temperature: Refrigerator
MDL Number: MFCD10699394
CAS Number: 205444-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR