2-Bromo-6-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

11047-25G 11047-5G
76894-500EA 240.83 USD
76894-500 76894-502
2-Bromo-6-iodopyridine ≥97%
2-Bromo-6-iodopyridine

Caution: for Research Use Only

Formula: C₅H₃BrIN
MW: 283.89
Storage Temperature: Ambient
MDL Number: MFCD08059557
CAS Number: 234111-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR