Dimethyl Sulfoxide, Procipient® USP, Ph. Eur., Gaylord Chemical

Supplier: Spectrum Chemical Mfg. Corp.
Warning

Synonyms: Methanesulfinylmethane|Methyl sulfoxide|Dimethyl(oxido)sulfur|DMSO|Methanesulfinylmethane

5010-450LT-CS
76894-072CS 8016.15 USD
76894-072
Dimethyl Sulfoxide, Procipient® USP, Ph. Eur., Gaylord Chemical
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
UN: 1993

Order Now

Specification Test Results

Assay, % by GC ≥99.99
Trademark Gaylord Chemical Company, LLC PROCIPIENT™ (Dimethyl Sulfoxide USP, PhEur) via Spectrum Chemical Mfg. Corp.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR