6-(Dimethylamino)nicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

23811-1G 23811-25G 23811-5G
76885-338EA 81.69 USD
76885-338 76885-340 76885-342
6-(Dimethylamino)nicotinic acid ≥97% research grade
6-(Dimethylamino)nicotinic acid
Formula: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07357408
CAS Number: 82846-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR