3-Bromo-2-fluorobenzylalkohol ≥97%

Supplier: AOBChem USA

Warning

21835-25G
76884-862EA 123.07 USD
76884-862
3-Bromo-2-fluorobenzylalkohol ≥97%
3-Bromo-2-fluorobenzylalkohol
Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD09842441
CAS Number: 261723-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR