2-Fluorophenetole ≥97%

Supplier: AOBChem USA

Warning

21089-25G
76884-698EA 112.46 USD
76884-698
2-Fluorophenetole ≥97%
2-Fluorophenetole

Caution: for Research Use Only

Formula: C₈H₉FO
Storage Temperature: Ambient
MDL Number: MFCD00017909
CAS Number: 451-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR