N-(3-Pyridyl)pivalamide ≥97% research grade

Supplier: AOBChem USA

Warning

20817-25G
76884-654EA 106.09 USD
76884-654
N-(3-Pyridyl)pivalamide ≥97% research grade
N-(3-Pyridyl)pivalamide
Formula: C₁₀H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00996243
CAS Number: 70298-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR