2,3,4-Trifluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3,4-Trifluorobenzeneboronic acid, Dihydroxy(2,3,4-trifluorophenyl)borane, (2,3,4-Trifluorophenyl)metaboric acid

10445-25G
76882-050EA 62.6 USD
76882-050
2,3,4-Trifluorophenylboronic acid ≥97%
2,3,4-Trifluorophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₄BF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01863168
CAS Number: 226396-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR