L(-)-Sorbose 95%

Supplier: Ambeed
Warning

Synonyms: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

A385279-10G A385279-25G A385279-100G
76876-724EA 37 USD
76876-724 76876-726 77884-020
L(-)-Sorbose 95%
L(-)-Sorbose

Caution: for Research Use Only

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 380
MDL Number: MFCD00151097
CAS Number: 87-79-6
Merck Index: 12,08874

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR