Mannose triflate 95%

Supplier: Ambeed
Danger

Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, TATM

A189531-100MG A189531-250MG A189531-5G A189531-1G
76873-008EA 22 USD
76873-008 76873-010 76873-012 77883-403
Mannose triflate 95%
Mannose triflate

Caution: for Research Use Only

Formula: C₁₅H₁₉F₃O₁₂S
MDL Number: MFCD00012353
CAS Number: 92051-23-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR