1-Bromo-4-[(trifluoromethyl)sulfonyl]benzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-((trifluoromethyl)sulphonyl)benzene, 1-Bromo-4-((trifluoromethyl)sulfonyl)benzene

A174144-1G A174144-250MG
76872-528EA 113 USD
76872-528 77883-336
1-Bromo-4-[(trifluoromethyl)sulfonyl]benzene 97%
1-Bromo-4-[(trifluoromethyl)sulfonyl]benzene

Caution: for Research Use Only

Formula: C₇H₄BrF₃O₂S
MDL Number: MFCD00474580
CAS Number: 312-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR