N-Boc-L-alaninol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol
A142574-1000G
A142574-100G
A142574-10G
A142574-25G
A142574-500G
A142574-5G
76871-280EA
354
USD
76871-280
76871-282
76871-284
76871-286
76871-288
76871-290
76871-292
N-Boc-L-alaninol 98%
N-Boc-L-alaninol
Caution: for Research Use Only
Formula:
C₈H₁₇NO₃ |
MDL Number:
MFCD00043121 CAS Number: 79069-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...