4-Chloro-3,5-xylenol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol

A139376-1000G A139376-100G A139376-25G A139376-500G
76871-130EA 55 USD
76871-130 76871-132 76871-134 76871-136 76871-138
4-Chloro-3,5-xylenol 98%
4-Chloro-3,5-xylenol

Caution: for Research Use Only

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246
Flash Pt: 132
MDL Number: MFCD00002324
CAS Number: 88-04-0
Merck Index: 13,02194

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR