3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A133455-1G
76870-866EA 121 USD
76870-866
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 97%
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₇H₂₅BFNO₃
MDL Number: MFCD09027076
CAS Number: 1073354-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR