Anti-PKC mu/PKD Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A11012-100
76865-388EA 458.7 USD
76865-388 76868-506
Anti-PKC mu/PKD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PKC mu / PKD (phospho Ser744 + Ser748) for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: PKC mu/PKD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR