Sampling Kit for Identifier System
Supplier: STAT PEEL INC
A00600
76848-022EA 5722.2 USD
76848-022
Sampling Kit for Identifier System
Samplers Air Samplers
The sampling kit allows for the collection of respirable particles for inspection in the STAT PEEL® Identifier system.
  • 6 Battery powered badges for respirable particle collection
  • Tablet computer for badge control
  • Badge chargers
  • All necessary tools to perform air samplings
  • Starter kit of 20 filtration slides
The sampling kit allows for the collection of respirable particles for inspection in the STAT PEEL® Identifier System or for SEM inspection. The included filtration badges are personal sampling devices equipped with a silent pump, environmental sensors, and filtration slides that collect respirable particles. It can be worn in a lab coat pocket to sample personal exposure or set up in a desired location to sample environmental contamination. The badge is operated with an easy-to-use app on a tablet computer. Up to 10 sampling badges can be controlled by one tablet. After completion of the sampling run, the badge data can be transferred to the tablet computer.
Certifications: CE marked.